Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: TNFSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266670.1 | NP_001253599.1 | 373 | 104 | ||
XM_015116969.2 | XP_014972455.1 | 599 | 104 | |||
GenBank mRNA | JU477903.1 | - | 373 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |