Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC696037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266719.2 | NP_001253648.1 | 227 | 79 | ||
XM_015146113.2 | XP_015001599.1 | 79 | ||||
GenBank mRNA | JU322979.1 | - | 330 | 79 | ||
JU471851.1 | - | 228 | 79 | |||
JV616749.1 | - | 79 |
Gene Symbol: LY6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028853026.1 | XP_028708859.1 | 116 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |