Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115825.2 | XP_014971311.2 | 3642 | 107 | ||
XM_015115826.2 | XP_014971312.2 | 3642 | 107 | |||
XM_028834035.1 | XP_028689868.1 | 3739 | 107 | |||
XM_028834036.1 | XP_028689869.1 | 1126 | 107 | |||
GenBank mRNA | JU343756.1 | - | 3206 | 107 | ||
JU486874.1 | - | 3218 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |