Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Brs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152845.1 | NP_690058.1 | 2 - 3 | 910 | 74 | |
XM_006257677.2 | XP_006257739.1 | 3 - 4 | 940 | 74 | ||
XM_006257678.2 | XP_006257740.1 | 3 - 4 | 759 | 74 | ||
GenBank mRNA | AF510984.1 | - | 2 - 3 | 910 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |