Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc26a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057127.1 | NP_476468.1 | 1 - 2 | 220 | 85 | |
XM_006254784.2 | XP_006254846.1 | 1 - 2 | 344 | 85 | ||
XM_006254785.2 | XP_006254847.1 | 1 - 2 | 344 | 85 | ||
XM_006254786.2 | XP_006254848.1 | 1 - 2 | 344 | 85 | ||
XM_006254787.2 | XP_006254849.1 | 1 - 2 | 344 | 85 | ||
GenBank mRNA | D82883.1 | - | 1 - 2 | 220 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |