Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019251.2 | NP_062124.1 | 1 - 2 | 102 | 79 | |
GenBank mRNA | BC062408.1 | - | 1 - 2 | 102 | 5621977 | 79 |
FQ214230.1 | - | 1 - 2 | 156 | 79 | ||
FQ223263.1 | - | 1 - 2 | 177 | 79 | ||
U42755.1 | - | 1 - 2 | 146 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |