Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402427_g1
          See other IL7R GE Assays ›
          Gene Symbol:
          Il7r
          Gene Name:
          interleukin 7 receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2|NW_007906130: 21860 - 47693 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402427_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il7r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106418.1 NP_001099888.1 7 - 8 955 106
          XM_006257698.2 XP_006257760.1 7 - 8 839 106
          XM_006257699.2 XP_006257761.1 7 - 8 813 106
          XM_008773706.1 XP_008771928.1 6 - 7 2751 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2_KL567908v1_random00000002.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          negative regulation of T cell mediated cytotoxicity
          immunoglobulin production
          biological_process
          regulation of cell size
          positive regulation of gene expression
          cell growth
          cytokine-mediated signaling pathway
          T cell differentiation
          positive regulation of T cell differentiation in thymus
          B cell proliferation
          lymph node development
          homeostasis of number of cells
          molecular_function
          cytokine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007943 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P828142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3110 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40413 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P828142 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007943 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P828142 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007943 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007943 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P828142 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P100584 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87554 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821854 - -
          Affymetrix Rat Gene 1.0 ST Array 10821857 - -
          Affymetrix Rat Gene 1.0 ST Array 10821856 - -
          Affymetrix Rat Gene 1.0 ST Array 10821858 - -
          Affymetrix Rat Gene 1.0 ST Array 10821852 - -
          Affymetrix Rat Gene 1.0 ST Array 10821859 - -
          Affymetrix Rat Gene 1.0 ST Array 10821853 - -
          Affymetrix Rat Gene 1.0 ST Array 10821855 - -
          Affymetrix Rat Gene 1.1 ST Array 10821851 - -
          Rat Genome 230 2.0 Array 1382566_at - -
          Rat Genome 230 2.0 Array 1386052_PM_at - -
          Rat Genome 230 2.0 Array 1382566_PM_at - -
          Rat Genome 230 2.0 Array 1386052_at - -
          Affymetrix HT Rat Focus Array 1382566_at - -
          Affymetrix HT Rat Focus Array 1386052_PM_at - -
          Affymetrix HT Rat Focus Array 1386052_at - -
          Affymetrix HT Rat Focus Array 1382566_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2_KL567908v1_random00000002.rn.1 TC2_KL567908v1_random00000002.rn.1 RTA TC
          Rat Expression Set 230 1386052_at - -
          Rat Expression Set 230 1386052_PM_at - -
          Rat Expression Set 230 1382566_at - -
          Rat Expression Set 230 1382566_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10821851 - -
          Rat Genome U34 Set rc_AA800754_at - -
          Rat Genome U34 Set rc_AI011994_at - -
          Rat Genome U34 Set rc_AI071569_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21993897 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358648 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline