Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: RGD735029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199493.1 | NP_955787.1 | 5 - 6 | 613 | 64 | |
XM_006254634.2 | XP_006254696.1 | 5 - 6 | 720 | 64 | ||
GenBank mRNA | BC064660.1 | - | 5 - 6 | 613 | 5623815 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |