Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf38a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106672.1 | NP_001100142.1 | 6 - 7 | 929 | 88 | |
GenBank mRNA | FQ221356.1 | - | 4 - 5 | 335 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |