Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01426944_g1
          See other CTU2 GE Assays ›
          Gene Symbol:
          Ctu2
          Gene Name:
          cytosolic thiouridylase subunit 2 homolog (S. pombe)
          Gene Aliases:
          Ncs2
          Chromosome Location:
          Chr.19: 55300531 - 55305543 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01426944_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctu2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037094.1 NP_001032171.1 12 - 13 1352 142
          GenBank mRNA BC079393.1 - 12 - 13 1402 142
          BC089862.1 - 12 - 13 1350 142
          BC107464.1 - 12 - 13 1352 7454431 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000778.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctu2

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA wobble uridine modification
          biological_process
          protein urmylation
          tRNA thio-modification
          tRNA binding
          molecular_function
          nucleotidyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531462 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029322 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029322 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029322 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P675672 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52177 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10808529 - -
          Affymetrix Rat Gene 1.0 ST Array 10808522 - -
          Affymetrix Rat Gene 1.0 ST Array 10808525 - -
          Affymetrix Rat Gene 1.0 ST Array 10808520 - -
          Affymetrix Rat Gene 1.0 ST Array 10808533 - -
          Affymetrix Rat Gene 1.0 ST Array 10808524 - -
          Affymetrix Rat Gene 1.0 ST Array 10808534 - -
          Affymetrix Rat Gene 1.0 ST Array 10808521 - -
          Affymetrix Rat Gene 1.0 ST Array 10808527 - -
          Affymetrix Rat Gene 1.0 ST Array 10808530 - -
          Affymetrix Rat Gene 1.0 ST Array 10808535 - -
          Affymetrix Rat Gene 1.0 ST Array 10808532 - -
          Affymetrix Rat Gene 1.0 ST Array 10808526 - -
          Affymetrix Rat Gene 1.0 ST Array 10808523 - -
          Affymetrix Rat Gene 1.0 ST Array 10808531 - -
          Affymetrix Rat Gene 1.1 ST Array 10808519 - -
          Rat Genome 230 2.0 Array 1372952_PM_at - -
          Rat Genome 230 2.0 Array 1372952_at - -
          Affymetrix HT Rat Focus Array 1372952_PM_at - -
          Affymetrix HT Rat Focus Array 1372952_at - -
          Affymetrix Rat Clariom S Assay PSR1900006301.rn.1 TC1900000778.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006300.rn.1 TC1900000778.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900006302.rn.1 TC1900000778.rn.1 RTA TC
          Rat Expression Set 230 1372952_PM_at - -
          Rat Expression Set 230 1372952_at - -
          Affymetrix Rat Gene 1.0 ST Array 10808519 - -
          Rat Genome U34 Set rc_AI071794_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353625 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline