Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153626.1 | NP_705890.1 | 5 - 6 | 852 | 130 | |
XM_006230697.2 | XP_006230759.1 | 7 - 8 | 3009 | 130 | ||
GenBank mRNA | AB050103.1 | - | 5 - 6 | 852 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |