Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ndufa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127684.2 | NP_001121156.1 | 1 - 2 | 164 | 58 | |
GenBank mRNA | BC167115.1 | - | 1 - 2 | 163 | 6922256 | 58 |
FM055502.1 | - | 1 - 2 | 60 | 58 | ||
FM088467.1 | - | 1 - 2 | 144 | 58 | ||
FQ210865.1 | - | 1 - 2 | 119 | 58 | ||
FQ217628.1 | - | 1 - 2 | 107 | 58 | ||
FQ224731.1 | - | 1 - 2 | 135 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |