Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Rai14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011947.1 | NP_001011947.1 | 15 - 16 | 2813 | 57 | |
XM_006232060.2 | XP_006232122.1 | 16 - 17 | 3387 | 57 | ||
XM_006232061.2 | XP_006232123.1 | 16 - 17 | 2960 | 57 | ||
XM_006232062.2 | XP_006232124.1 | 16 - 17 | 3825 | 57 | ||
XM_006232063.2 | XP_006232125.1 | 15 - 16 | 3300 | 57 | ||
XM_006232064.2 | XP_006232126.1 | 14 - 15 | 3213 | 57 | ||
XM_008760765.1 | XP_008758987.1 | 14 - 15 | 3756 | 57 | ||
GenBank mRNA | BC085775.1 | - | 15 - 16 | 2813 | 7096978 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |