Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108071.1 | NP_001101541.1 | 4 - 5 | 445 | 153 | |
XM_006240915.2 | XP_006240977.1 | 6 - 7 | 1374 | 153 | ||
XM_006240916.2 | XP_006240978.1 | 6 - 7 | 1364 | 153 | ||
XM_006240917.2 | XP_006240979.1 | 6 - 7 | 1371 | 153 | ||
XM_006240918.2 | XP_006240980.1 | 6 - 7 | 1314 | 153 | ||
XM_006240919.2 | XP_006240981.1 | 6 - 7 | 1373 | 153 | ||
XM_006240920.2 | XP_006240982.1 | 5 - 6 | 1203 | 153 | ||
GenBank mRNA | FQ224249.1 | - | 6 - 7 | 674 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |