Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sele | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138879.1 | NP_620234.1 | 10 - 11 | 1589 | 141 | |
XM_008769608.1 | XP_008767830.1 | 12 - 13 | 2160 | 141 | ||
GenBank mRNA | L25527.1 | - | 10 - 11 | 1589 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |