Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008286.1 | NP_001008287.1 | 1 - 2 | 204 | 84 | |
XM_006234371.2 | XP_006234433.1 | 1 - 2 | 237 | 84 | ||
GenBank mRNA | BC086360.1 | - | 1 - 2 | 204 | 7303884 | 84 |
FQ213350.1 | - | 1 - 2 | 178 | 84 | ||
FQ215122.1 | - | 1 - 2 | 172 | 84 | ||
FQ215507.1 | - | 1 - 2 | 199 | 84 | ||
FQ215582.1 | - | 1 - 2 | 222 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |