Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191828.1 | NP_001178757.1 | 11 - 12 | 1683 | 73 | |
XM_006250175.2 | XP_006250237.1 | 9 - 10 | 1140 | 73 | ||
XM_006250176.2 | XP_006250238.1 | 10 - 11 | 1165 | 73 | ||
XM_008769660.1 | XP_008767882.1 | 10 - 11 | 2509 | 73 | ||
XM_008769661.1 | XP_008767883.1 | 10 - 11 | 2509 | 73 | ||
XM_008769662.1 | XP_008767884.1 | 11 - 12 | 1567 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |