Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106780.1 | NP_001100250.1 | 1 - 2 | 227 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |