Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lgi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199499.1 | NP_955793.1 | 7 - 8 | 1001 | 65 | |
XM_008759169.1 | XP_008757391.1 | 6 - 7 | 1444 | 65 | ||
XM_008759170.1 | XP_008757392.1 | 7 - 8 | 715 | 65 | ||
GenBank mRNA | BC064659.1 | - | 7 - 8 | 1001 | 5623799 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |