Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008814.1 | NP_001008814.1 | 7 - 8 | 1248 | 139 | |
XM_008765774.1 | XP_008763996.1 | 7 - 8 | 1294 | 139 | ||
GenBank mRNA | AB355638.1 | - | 7 - 8 | 1248 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |