Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RPL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001180895.1 | NP_011485.1 | 1 | 252 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Endogenous Controls Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |