Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348770.1 | NP_001335699.1 | 2 | 376 | 93 | |
NR_145981.2 | 2 | 376 | 93 | |||
GenBank mRNA | AB116559.1 | - | 2 | 335 | 93 | |
AK238831.1 | - | 2 | 359 | 93 | ||
AK239307.1 | - | 1 | 441 | 93 | ||
AK239664.1 | - | 2 | 357 | 93 | ||
AK398365.1 | - | 2 | 365 | 93 | ||
AY438642.1 | - | 2 | 354 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |