Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244093.1 | NP_001231022.1 | 5 - 6 | 649 | 52 | |
XM_013995256.2 | XP_013850710.1 | 1180 | 52 | |||
GenBank mRNA | AK349993.1 | - | 5 - 6 | 640 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |