Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: matn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030442.1 | NP_001025613.1 | 6 - 7 | 1349 | 77 | |
XM_018091297.1 | XP_017946786.1 | 6 - 7 | 1382 | 77 | ||
GenBank mRNA | BC091071.1 | - | 6 - 7 | 1349 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |