Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: syf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037263.1 | NP_001032340.1 | 1 - 2 | 57 | 68 | |
GenBank mRNA | BC087564.2 | - | 1 - 2 | 39 | 68 | |
BC122018.1 | - | 1 - 2 | 52 | 68 | ||
BC159306.1 | - | 1 - 2 | 76 | 68 | ||
CR761644.2 | - | 1 - 2 | 57 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |