Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045689.2 | NP_001039154.1 | 10 - 11 | 1268 | 82 | |
GenBank mRNA | CR942586.2 | - | 9 - 10 | 1197 | 82 | |
DR897520.1 | - | 1 - 2 | 386 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |