Search Thermo Fisher Scientific
Gene Symbol: NAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130460.1 | NP_001123932.1 | 750 | 97 | ||
GenBank mRNA | EU963406.1 | - | 959 | 97 | ||
JQ217429.1 | - | 750 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Zea mays |