Search
Search

AT4G21930| Gene Symbol: AT4G21930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118314.3 | NP_193924.2 | 1 | 555 | 69 | |
| GenBank mRNA | AY120720.1 | - | 1 | 555 | 69 | |
| BT000027.1 | - | 1 | 419 | 69 | ||
Set Membership: |
|