Search
Search

AT2G16790| Gene Symbol: AT2G16790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124859.1 | NP_001118331.1 | 4 - 5 | 247 | 95 | |
| NM_001335490.1 | NP_001325318.1 | 3 - 4 | 396 | 95 | ||
| NM_001335492.1 | NP_001325317.1 | 3 - 4 | 428 | 95 | ||
| NM_127231.2 | NP_565393.1 | 4 - 5 | 361 | 95 | ||
Set Membership: |
|