Search
Search

DXPS1| Gene Symbol: DXPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338537.1 | NP_001327775.1 | 7 - 8 | 1731 | 71 | |
| NM_113045.3 | NP_566686.2 | 7 - 8 | 1630 | 71 | ||
| NM_180289.4 | NP_850620.2 | 7 - 8 | 1731 | 71 | ||
Set Membership: |
|