Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BTR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120525.4 | NP_196063.1 | 7 - 8 | 1068 | 103 | |
NM_180433.3 | NP_850764.1 | 7 - 8 | 1131 | 103 | ||
GenBank mRNA | AF375411.1 | - | 7 - 8 | 930 | 103 | |
AY086553.1 | - | 7 - 8 | 948 | 103 | ||
BT000854.1 | - | 6 - 7 | 779 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |