Search
Search

AT5G07890| Gene Symbol: AT5G07890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036773.2 | NP_001031850.1 | 5 - 6 | 1243 | 76 | |
| NM_001036774.2 | NP_001031851.1 | 7 - 8 | 1226 | 76 | ||
| NM_001342965.1 | NP_001318504.1 | 7 - 8 | 1214 | 76 | ||
| NM_001342966.1 | NP_001332408.1 | 6 - 7 | 1389 | 76 | ||
| NM_120871.3 | NP_196406.2 | 6 - 7 | 1145 | 76 | ||
| GenBank mRNA | AK317060.1 | - | 6 - 7 | 1149 | 76 | |
Set Membership: |
|