Search
Search

YLS8| Gene Symbol: YLS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120912.3 | NP_196446.1 | 1 - 2 | 262 | 58 | |
| GenBank mRNA | AB047811.1 | - | 1 - 2 | 153 | 58 | |
| AF324685.2 | - | 1 - 2 | 183 | 58 | ||
| AF326857.1 | - | 1 - 2 | 183 | 58 | ||
| AF339680.1 | - | 1 - 2 | 118 | 58 | ||
| AF375407.1 | - | 1 - 2 | 183 | 58 | ||
| AY066051.1 | - | 1 - 2 | 118 | 58 | ||
Set Membership: |
|