Search
Search

AT3G52155| Gene Symbol: AT3G52155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339557.1 | NP_001325732.1 | 2 - 3 | 493 | 92 | |
| NM_001339558.1 | NP_001325733.1 | 2 - 3 | 335 | 92 | ||
| NM_001339559.1 | NP_001325731.1 | 1 - 2 | 160 | 92 | ||
| NM_115075.3 | NP_566959.1 | 2 - 3 | 493 | 92 | ||
| GenBank mRNA | AY039568.1 | - | 2 - 3 | 335 | 92 | |
Set Membership: |
|