Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084826.2 | NP_001078295.1 | 4 - 5 | 996 | 116 | |
NM_115398.3 | NP_191100.1 | 4 - 5 | 1028 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |