Search
Search

ROPGEF6| Gene Symbol: ROPGEF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339743.1 | NP_001325478.1 | 5 - 6 | 1099 | 156 | |
| NM_115424.3 | NP_191125.1 | 5 - 6 | 1140 | 156 | ||
| GenBank mRNA | BT020279.1 | - | 5 - 6 | 1023 | 156 | |
| BT021117.1 | - | 5 - 6 | 946 | 156 | ||
Set Membership: |
|