Search
Search

AT3G60510| Gene Symbol: AT3G60510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203215.1 | NP_001190144.1 | 10 - 11 | 1137 | 74 | |
| NM_115915.5 | NP_191610.3 | 11 - 12 | 1161 | 74 | ||
| GenBank mRNA | BT015757.1 | - | 11 - 12 | 1012 | 74 | |
| BT020182.1 | - | 11 - 12 | 947 | 74 | ||
Set Membership: |
|