Search
Search

GATA4| Gene Symbol: GATA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115917.5 | NP_191612.1 | 1 - 2 | 544 | 69 | |
| GenBank mRNA | AF378881.1 | - | 1 - 2 | 239 | 69 | |
| AY039532.1 | - | 1 - 2 | 239 | 69 | ||
| AY050476.1 | - | 1 - 2 | 209 | 69 | ||
| Y13651.1 | - | 214 | 69 | |||
Set Membership: |
|