Search
Search

AT5G14130| Gene Symbol: AT5G14130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343316.1 | NP_001332223.1 | 1 - 2 | 517 | 83 | |
| NM_121417.5 | NP_196917.1 | 1 - 2 | 339 | 83 | ||
| GenBank mRNA | AY057607.1 | - | 1 - 2 | 269 | 83 | |
| AY124834.1 | - | 1 - 2 | 230 | 83 | ||
| X98806.1 | - | 258 | 83 | |||
Set Membership: |
|