Search
Search

TERT| Gene Symbol: TERT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343468.1 | NP_001330126.1 | 5 - 6 | 1114 | 120 | |
| NM_121691.4 | NP_197187.1 | 5 - 6 | 1016 | 120 | ||
| GenBank mRNA | AF135454.1 | - | 5 - 6 | 1000 | 120 | |
Set Membership: |
|