Search Thermo Fisher Scientific
Gene Symbol: ABCB21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340142.1 | NP_001327193.1 | 1 - 2 | 704 | 104 | |
NM_001340143.1 | NP_001327192.1 | 2 - 3 | 729 | 104 | ||
NM_116080.3 | NP_191774.2 | 2 - 3 | 747 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |