Search
Search

AT3G62500| Gene Symbol: AT3G62500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340158.1 | NP_001326690.1 | 5 - 6 | 961 | 74 | |
| NM_116115.1 | NP_191809.1 | 5 - 6 | 961 | 74 | ||
Set Membership: |
|