Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: AGD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340549.1 | NP_001328044.1 | 4 - 5 | 342 | 122 | |
NM_116772.4 | NP_567292.1 | 4 - 5 | 388 | 122 | ||
GenBank mRNA | AY084733.1 | - | 4 - 5 | 258 | 122 | |
BT024733.1 | - | 3 - 4 | 199 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |