Search
Search

DDB1A| Gene Symbol: DDB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125468.1 | NP_001118940.1 | 15 - 16 | 2150 | 127 | |
| NM_116781.3 | NP_192451.1 | 16 - 17 | 2221 | 127 | ||
| GenBank mRNA | BT001905.1 | - | 16 - 17 | 2047 | 127 | |
Set Membership: |
|