Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TLP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104210.4 | NP_564627.1 | 2 - 3 | 480 | 122 | |
GenBank mRNA | AF370146.1 | - | 2 - 3 | 467 | 122 | |
AY059088.1 | - | 2 - 3 | 405 | 122 | ||
AY092403.1 | - | 2 - 3 | 405 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |