Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZCF61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333855.1 | NP_001320817.1 | 2 - 3 | 320 | 89 | |
NM_001333856.1 | NP_001320816.1 | 2 - 3 | 278 | 89 | ||
NM_001333857.1 | NP_001320818.1 | 2 - 3 | 255 | 89 | ||
NM_104650.4 | NP_564745.1 | 2 - 3 | 320 | 89 | ||
GenBank mRNA | AK228076.1 | - | 2 - 3 | 255 | 89 | |
BT006461.1 | - | 2 - 3 | 113 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |