Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› At02214607_g1
See other PEN3 GE Assays ›
Gene Symbol:
PEN3
Gene Name:
ABC-2 and Plant PDR ABC-type transporter family protein
Gene Aliases:
ABCG36, ARABIDOPSIS PLEIOTROPIC DRUG RESISTANCE 8, ARABIDOPSIS THALIANA ATP-BINDING CASSETTE G36, AT1G59870, ATABCG36, ATP-BINDING CASSETTE G36, ATP-binding cassette G36, ATPDR8, Arabidopsis thaliana ATP-binding cassette G36, F23H11.19, F23H11_19, PDR8, PENETRATION 3, PLEIOTROPIC DRUG RESISTANCE 8
Chromosome Location:
Chr.1: 22034423 - 22040126 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID
At02214607_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PEN3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_104680.3
NP_176196.1
7 - 8
3725
106
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Detect Genomic DNA
Recommended Assay
Arabidopsis thaliana
Probe spans exons
Panther Classification:
Molecular Function -
ATP-binding cassette (ABC) transporter
Gene Ontology Categories:
Function(s)
Process(es)
defense response to oomycetes
response to molecule of bacterial origin
response to molecule of fungal origin
response to oomycetes
response to molecule of oomycetes origin
response to water deprivation
response to bacterium
response to fungus
plant-type hypersensitive response
systemic acquired resistance
response to salt stress
response to auxin
response to abscisic acid
coumarin metabolic process
defense response to fungus, incompatible interaction
auxin polar transport
response to chitin
regulation of auxin mediated signaling pathway
cadmium ion transport
negative regulation of defense response
indole glucosinolate catabolic process
defense response to bacterium
thymidine metabolic process
root development
cotyledon development
defense response to fungus
defense response by callose deposition in cell wall
sodium ion homeostasis
transmembrane transport
cellular response to indolebutyric acid stimulus
root hair cell development
cellular detoxification
regulation of lateral root development
regulation of defense response by callose deposition
mRNA binding
protein binding
auxin efflux transmembrane transporter activity
cadmium ion transmembrane transporter activity
efflux transmembrane transporter activity
ATPase activity
ATPase activity, coupled to transmembrane movement of substances
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0