Search
Search

ABCF3| Gene Symbol: ABCF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105130.4 | NP_176636.1 | 2 - 3 | 518 | 91 | |
| GenBank mRNA | BT002015.1 | - | 2 - 3 | 460 | 91 | |
| BT010544.1 | - | 2 - 3 | 418 | 91 | ||
Set Membership: |
|