Search
Search

G-TMT| Gene Symbol: G-TMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105171.3 | NP_176677.1 | 2 - 3 | 643 | 53 | |
| GenBank mRNA | AY049258.1 | - | 2 - 3 | 594 | 53 | |
| AY090280.1 | - | 2 - 3 | 527 | 53 | ||
Set Membership: |
|