Search
Search

NUDT7| Gene Symbol: NUDT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203778.1 | NP_001190707.1 | 8 - 9 | 958 | 123 | |
| NM_001340798.1 | NP_001329574.1 | 7 - 8 | 891 | 123 | ||
| NM_001340799.1 | NP_001329573.1 | 5 - 6 | 457 | 123 | ||
| NM_117341.5 | NP_193008.1 | 8 - 9 | 748 | 123 | ||
| NM_179036.4 | NP_849367.1 | 7 - 8 | 914 | 123 | ||
| NM_179037.3 | NP_849368.1 | 7 - 8 | 586 | 123 | ||
| GenBank mRNA | AF325104.1 | - | 6 - 7 | 514 | 123 | |
| AF370209.1 | - | 7 - 8 | 572 | 123 | ||
| AY056344.1 | - | 6 - 7 | 514 | 123 | ||
Set Membership: |
|